Learn More
Sigma Aldrich Fine Chemicals Biosciences 1,3-Dioxolane ReagentPlus(R), contains ~50 ppm BHT as inhibitor, 99% | 646-06-0 | MFCD00003207 | 4L
Catalog No. 501883975
Encompass
1,3-Dioxolane ReagentPlus(R), contains ~50 ppm BHT as inhibitor, 99% | Purity: 99% | Mol Wt: 74.08 | 646-06-0 | MFCD00003207 | 4L
Catalog No. 50-188-3975 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1844974L
1,3-Dioxolane ReagentPlus(R), contains ~50 ppm BHT as inhibitor, 99% | Purity: 99% | Mol Wt: 74.08 | 646-06-0 | MFCD00003207 | 4L