Learn More
Medchemexpress LLC 1,3-dipalmitoyl-glycero-2-phosphoethanolamine | 67303-93-9 | MFCD00038196 | >98.0% | 691.96 | C37H74NO8P | 50 MG
SDP
Catalog No. 5000282035
Encompass_Preferred
1,3-Dipalmitoyl-glycero-2-phosphoethanolamine (1,3-DPPE) is a phosphoethanolamine-class lipid used as a research reagent and building block for liposome formulation, membrane modelling, and biophysical studies. The molecule contains two palmitoyl (C16) acyl chains and a phosphoethanolamine headgroup, giving it saturated-chain bilayer properties and suitability for stable membrane systems.
  • Saturated phosphoethanolamine lipid with two C16 (palmitoyl) chains.
  • Suitable for liposome and membrane model formulation.
  • Promotes stable bilayer formation and higher phase transition temperature.
  • High chemical purity suitable for analytical and preparative use.
  • Powder format with recommended cold storage for extended stability.
Catalog No. 50-002-82035 Supplier Medchemexpress LLC Supplier No. HYW34373650MG
1,3-Dipalmitoyl-glycero-2-phosphoethanolamine (1,3-DPPE) is a phosphoethanolamine-class lipid used as a research reagent and building block for liposome formulation, membrane modelling, and biophysical studies. The molecule contains two palmitoyl (C16) acyl chains and a phosphoethanolamine headgroup, giving it saturated-chain bilayer properties and suitability for stable membrane systems.
  • Saturated phosphoethanolamine lipid with two C16 (palmitoyl) chains.
  • Suitable for liposome and membrane model formulation.
  • Promotes stable bilayer formation and higher phase transition temperature.
  • High chemical purity suitable for analytical and preparative use.
  • Powder format with recommended cold storage for extended stability.