Learn More
Medchemexpress LLC 1,3-dipalmitoyl-glycero-2-phosphoethanolamine | 67303-93-9 | MFCD00038196 | 99.0% | 691.96 g/mol | C37H74NO8P | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW34373625MG
Encompass_Preferred
1,3-Dipalmitoyl-glycero-2-phosphoethanolamine is a synthetic phospholipid used to prepare model membranes, liposomes, and micelles for biochemical and biophysical research. With two saturated palmitoyl (16:0) acyl chains and a phosphoethanolamine headgroup, it supports formation of ordered bilayer structures and is suitable for membrane studies and formulation work.
  • Supports formation of stable liposomes and bilayers.
  • High chemical purity suitable for analytical and preparative work.
  • Long saturated acyl chains promote ordered membrane phases.
  • Soluble in common organic solvents for lipid film preparation.
  • Stable under recommended frozen storage as powder or in solution.
Catalog No. 50-002-80926