Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-diphenylurea-d10 | 108009-46-7 | 99.0% | 222.31 | C13H2D10N2O | 1 MG
SDP

Catalog No. 5000304016
Encompass_Preferred

1,3-diphenylurea-d10 is the deuterium-labeled (d10) isotopologue of 1,3-diphenylurea, supplied as a high-purity reference standard for analytical and research use. It is intended for use as an internal standard or tracer in mass spectrometry and other isotopic labeling applications.

  • High purity (99.0%) suitable for analytical standards.
  • Deuterium-labeled isotopologue for use as an internal or tracer standard.
  • Available in small milligram pack sizes for research workflows.
  • Molecular weight 222.31 and formula C13H2D10N2O for reference.
  • Documentation available: COA, SDS, and handling instructions.
  • Storage guidance provided for room temperature and frozen solvent conditions.

Catalog No. 50-003-04016 Supplier Medchemexpress LLC Supplier No. HYW014286S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-diphenylurea-d10 is the deuterium-labeled (d10) isotopologue of 1,3-diphenylurea, supplied as a high-purity reference standard for analytical and research use. It is intended for use as an internal standard or tracer in mass spectrometry and other isotopic labeling applications.

  • High purity (99.0%) suitable for analytical standards.
  • Deuterium-labeled isotopologue for use as an internal or tracer standard.
  • Available in small milligram pack sizes for research workflows.
  • Molecular weight 222.31 and formula C13H2D10N2O for reference.
  • Documentation available: COA, SDS, and handling instructions.
  • Storage guidance provided for room temperature and frozen solvent conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.