Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-diphenylurea-d10 | 108009-46-7 | 99.0% | 222.31 g·mol-1 | C13H2D10N2O | 10 MG
SDP

Catalog No. 5000302809
Encompass_Preferred

1,3-Diphenylurea-d10 is a deuterium-labeled analogue of 1,3-diphenylurea used as a stable isotope reagent and internal standard in analytical and tracer studies. Supplied in small research quantities, it is intended for use in mass spectrometry method development, quantitative analysis, and mechanistic experiments requiring isotopic labeling.

  • Deuterium-labeled internal standard for mass spectrometry and tracer studies.
  • High purity (99.0%) suitable for quantitative analysis.
  • Molecular weight approximately 222.31 g·mol-1; formula C13H2D10N2O.
  • Supplied in small research pack sizes for analytical workflows.
  • Defined storage recommendations for neat and solvent conditions to preserve stability.

Catalog No. 50-003-02809 Supplier Medchemexpress LLC Supplier No. HYW014286S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Diphenylurea-d10 is a deuterium-labeled analogue of 1,3-diphenylurea used as a stable isotope reagent and internal standard in analytical and tracer studies. Supplied in small research quantities, it is intended for use in mass spectrometry method development, quantitative analysis, and mechanistic experiments requiring isotopic labeling.

  • Deuterium-labeled internal standard for mass spectrometry and tracer studies.
  • High purity (99.0%) suitable for quantitative analysis.
  • Molecular weight approximately 222.31 g·mol-1; formula C13H2D10N2O.
  • Supplied in small research pack sizes for analytical workflows.
  • Defined storage recommendations for neat and solvent conditions to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.