Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dipropoxybenzene 98.0+%, TCI America™
SDP

Catalog No. D13621ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13621ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D13621ML Supplier TCI America Supplier No. D13621ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56106-37-7
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.27
MDL Number MFCD00059402
InChI Key UTFRNSPYRPYKDV-UHFFFAOYSA-N
Synonym Resorcinol Dipropyl Ether
PubChem CID 573684
IUPAC Name 1,3-dipropoxybenzene
SMILES CCCOC1=CC(OCCC)=CC=C1

Specifications

Color Yellow
Boiling Point 251°C
Quantity 1 mL
Formula Weight 194.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dipropoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.