Learn More
Medchemexpress LLC 1,3-distearoyl-2-chloropropanediol | 1329796-49-7 | 99.1% | 648.49 g/mol | C39H70D5ClO4 | 10 MG
SDP
Catalog No. 5000284681
Encompass_Preferred
1,3-Distearoyl-2-chloropropanediol is a fatty-acid ester used as an analytical standard and reagent in lipid research, method development, and quantitative analyses. It is supplied in small milligram quantities and is available in non-deuterated and deuterated forms for isotope-labeling workflows.
  • High purity for analytical and reference use.
  • Available in small milligram quantities for method development.
  • Deuterated variants available for isotope-labeling and quantification.
  • Suitable for mass spectrometry and chromatographic analyses of lipids.
  • Stable solid form for convenient handling and storage.
  • Documentation available including COA and SDS to support quality control.
Catalog No. 50-002-84681 Supplier Medchemexpress LLC Supplier No. HYW58580910MG
1,3-Distearoyl-2-chloropropanediol is a fatty-acid ester used as an analytical standard and reagent in lipid research, method development, and quantitative analyses. It is supplied in small milligram quantities and is available in non-deuterated and deuterated forms for isotope-labeling workflows.
  • High purity for analytical and reference use.
  • Available in small milligram quantities for method development.
  • Deuterated variants available for isotope-labeling and quantification.
  • Suitable for mass spectrometry and chromatographic analyses of lipids.
  • Stable solid form for convenient handling and storage.
  • Documentation available including COA and SDS to support quality control.