Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-distearoyl-2-chloropropanediol | 26787-56-4 | >99.0% | 643.46 g·mol⁻¹ | C39H75ClO4 | 5 MG
SDP

Catalog No. 5000284544
Encompass_Preferred

1,3-Distearoyl-2-chloropropanediol is a stearic-acid ester lipid reagent provided for laboratory research as an analytical standard. The compound is supplied in small milligram quantities with supporting documentation for analytical use, and labeled variants are available for specialized studies.

  • High chemical purity: >99.0%.
  • Molecular formula: C39H75ClO4.
  • Molecular weight: 643.46 g·mol⁻¹.
  • Available in small mg package sizes (example: 5 mg).
  • Isotopically labeled variants available for analytical applications.
  • Datasheet, certificate of analysis, and safety data sheet are provided.

Catalog No. 50-002-84544 Supplier Medchemexpress LLC Supplier No. HYW5858095MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Distearoyl-2-chloropropanediol is a stearic-acid ester lipid reagent provided for laboratory research as an analytical standard. The compound is supplied in small milligram quantities with supporting documentation for analytical use, and labeled variants are available for specialized studies.

  • High chemical purity: >99.0%.
  • Molecular formula: C39H75ClO4.
  • Molecular weight: 643.46 g·mol⁻¹.
  • Available in small mg package sizes (example: 5 mg).
  • Isotopically labeled variants available for analytical applications.
  • Datasheet, certificate of analysis, and safety data sheet are provided.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.