Learn More
Cayman Chemical 1,3-Distearoyl Glycerol, Purity: ≥98%, CAS: 504-40-5, 100 mg
SDP
Catalog No. 502937665
Encompass_Preferred
A diacylglycerol; has been found in wheat bran extract; has been used in the synthesis of prodrug forms of the NSAIDs ibuprofen, naproxen, and diclofenac that have reduced ulcerogenicity
Catalog No. 50-293-7665 Supplier Cayman Chemical Supplier No. 26946100 MG
A diacylglycerol; has been found in wheat bran extract; has been used in the synthesis of prodrug forms of the NSAIDs ibuprofen, naproxen, and diclofenac that have reduced ulcerogenicity