Learn More
Cayman Chemical 1,3-Distearoyl Glycerol, Purity: ≥98%, CAS: 504-40-5, 25 mg
SDP
Catalog No. 502938342
Encompass_Preferred
A diacylglycerol; has been found in wheat bran extract; has been used in the synthesis of prodrug forms of the NSAIDs ibuprofen, naproxen, and diclofenac that have reduced ulcerogenicity
Catalog No. 50-293-8342 Supplier Cayman Chemical Supplier No. 2694625 MG
A diacylglycerol; has been found in wheat bran extract; has been used in the synthesis of prodrug forms of the NSAIDs ibuprofen, naproxen, and diclofenac that have reduced ulcerogenicity