Learn More
Medchemexpress LLC 1,3-disubstituted-7-aryl-5,5-bis(trifluoromethyl)-5,8-dihydropyrimido[4,5-d]pyrimidine-2,4-dione | 488097-06-9 | 99.7% | 440.73 | C16H11ClF6N4O2 | 50 MG
SDP
Catalog No. 5000211335
Encompass_Preferred
GLP-1R Antagonist 1 (compound 5d) is an orally active, CNS-penetrant, non-competitive antagonist of the glucagon-like peptide-1 receptor (GLP-1R). It is used in preclinical biochemical and pharmacological research to study GLP-1R signaling and receptor pharmacology. The compound is supported by supplier analytical data and safety documentation for laboratory use.
  • Orally active and CNS penetrant.
  • Non-competitive antagonist of GLP-1R.
  • Documented in vitro activity (IC50: 650 nM).
  • Provided with analytical data and safety documentation.
  • Available in small research quantities, including 50 mg.
Catalog No. 50-002-11335 Supplier Medchemexpress LLC Supplier No. HY10111650MG
GLP-1R Antagonist 1 (compound 5d) is an orally active, CNS-penetrant, non-competitive antagonist of the glucagon-like peptide-1 receptor (GLP-1R). It is used in preclinical biochemical and pharmacological research to study GLP-1R signaling and receptor pharmacology. The compound is supported by supplier analytical data and safety documentation for laboratory use.
  • Orally active and CNS penetrant.
  • Non-competitive antagonist of GLP-1R.
  • Documented in vitro activity (IC50: 650 nM).
  • Provided with analytical data and safety documentation.
  • Available in small research quantities, including 50 mg.