Learn More
Apexbio Technology LLC 1-3-DITHIANE-500MG
SDP
Catalog No. 5000568787
Encompass_Preferred
1 3-Dithiane (CAS No 505-23-7) is a sulfur-containing organic compound often utilized in biomedical research as a protective group for aldehydes and ketones Originating from thioacetal chemistry it is integral in synthetic organic chemistry modulating carbonyl reactivity Its mechanism involves the stabilization of reactive intermediates in various organic reactions In vitro activity typically measures in the nanomolar to low micromolar range demonstrating its efficacy in precise reaction control Common applications include cell model studies animal research and drug screening with experimental concentrations dependent on specific research designs
Catalog No. 50-005-68787 Supplier Apexbio Technology LLC Supplier No. C6599500
1 3-Dithiane (CAS No 505-23-7) is a sulfur-containing organic compound often utilized in biomedical research as a protective group for aldehydes and ketones Originating from thioacetal chemistry it is integral in synthetic organic chemistry modulating carbonyl reactivity Its mechanism involves the stabilization of reactive intermediates in various organic reactions In vitro activity typically measures in the nanomolar to low micromolar range demonstrating its efficacy in precise reaction control Common applications include cell model studies animal research and drug screening with experimental concentrations dependent on specific research designs