Learn More
Medchemexpress LLC 1,3-Dithiane | 505-23-7 | 120.24 | 25 G
SDP
Catalog No. 5000449918
Encompass_Preferred
1,3-Dithiane is a protected formaldehyde anion equivalent and a sulfur-containing Maillard reaction product. It is found in cooked beef extracts and is a potent mutagenic agent against *S. typhimurium* TA98 and TA100. It can be used as a useful marker synthon.
  • Functions as a protected formaldehyde anion equivalent
  • Sulfur-containing Maillard reaction product
  • Identified in cooked beef extracts
  • Exhibits potent direct mutagenicity
  • Induces sister chromatid exchange (SCE) frequency
  • Utilized as a useful marker synthon
Catalog No. 50-004-49918 Supplier Medchemexpress LLC Supplier No. 25GHYW001189
1,3-Dithiane is a protected formaldehyde anion equivalent and a sulfur-containing Maillard reaction product. It is found in cooked beef extracts and is a potent mutagenic agent against *S. typhimurium* TA98 and TA100. It can be used as a useful marker synthon.
  • Functions as a protected formaldehyde anion equivalent
  • Sulfur-containing Maillard reaction product
  • Identified in cooked beef extracts
  • Exhibits potent direct mutagenicity
  • Induces sister chromatid exchange (SCE) frequency
  • Utilized as a useful marker synthon