Learn More
1,3-Dithiane, 98%
Description
1,3-Dithiane is used in umpolung reactions such as the Corey-Seebach reaction. It acts as a reagent for the deoxygenation of sulfoxides to their corresponding sulfides. Further, it is used as a protecting group for carbonyl compounds used in organic synthesis. It serves as a useful labeled synthon. In addition to this, it is used in the preparation of 2-ethyl-[1,3]dithiane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,3-Dithiane is used in umpolung reactions such as the Corey-Seebach reaction. It acts as a reagent for the deoxygenation of sulfoxides to their corresponding sulfides. Further, it is used as a protecting group for carbonyl compounds used in organic synthesis. It serves as a useful labeled synthon. In addition to this, it is used in the preparation of 2-ethyl-[1,3]dithiane.
Solubility
Soluble in benzene, ether, chloroform and terahydrofuran. Slightly soluble in water.
Notes
Hygroscopic. Incompatible with strong bases and strong oxidizing agents.
Specifications
Specifications
| Melting Point | 52°C to 55°C |
| Boiling Point | 207°C to 208°C |
| Flash Point | 90°C (194°F) |
| Odor | Stench |
| Quantity | 25 g |
| UN Number | UN3335 |
| Beilstein | 102534 |
| Solubility Information | Soluble in benzene,ether,chloroform and terahydrofuran. Slightly soluble in water. |
| Formula Weight | 120.24 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S. |
| EINECSNumber | 208-006-7 |
| RTECSNumber | JO5070000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.