Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dithiole-2-thione 97.0+%, TCI America™
SDP

Catalog No. D21335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21335G 5 g
D21331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21335G Supplier TCI America Supplier No. D21335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 930-35-8
Molecular Formula C3H2S3
Molecular Weight (g/mol) 134.23
MDL Number MFCD00014536
InChI Key WYKJWNVWJOKVQP-UHFFFAOYSA-N
PubChem CID 70254
IUPAC Name 2H-1,3-dithiole-2-thione
SMILES S=C1SC=CS1

Specifications

Melting Point 49°C
Quantity 5 g
Formula Weight 134.23
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dithiole-2-thione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.