Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethanamine | 253168-94-4 | 99.96% | 273.35 | 1 KG
SDP

Catalog No. 5000429706
Encompass_Preferred

1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethanamine is a drug intermediate for the synthesis of various active compounds. It is for research use only.

  • Purity: 99.96%
  • Molecular weight: 273.35
  • Formula: C12H19NO4S
  • Cas no.: 253168-94-4
  • Appearance: Solid

Catalog No. 50-004-29706 Supplier Medchemexpress LLC Supplier No. HYI06381KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethanamine is a drug intermediate for the synthesis of various active compounds. It is for research use only.

  • Purity: 99.96%
  • Molecular weight: 273.35
  • Formula: C12H19NO4S
  • Cas no.: 253168-94-4
  • Appearance: Solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.