Learn More
Medchemexpress LLC 1-(3-fluorophenyl)-2-((2-((2,3,4-trifluorophenyl)methoxy)phenyl)methyl)pyrrolidine hydrochloride | 1227056-87-2 | 99.5% | 357.80 g/mol | C18H19ClF3NO | 5 MG
SDP
Catalog No. 5000239920
Encompass_Preferred
Ampreloxetine hydrochloride is an orally active, CNS-penetrant inhibitor of the norepinephrine transporter (NET) and the serotonin uptake transporter (SERT) supplied for research use. It is used in vitro and in vivo to investigate monoamine transporter pharmacology and CNS transporter-related mechanisms.
  • Orally active inhibitor of norepinephrine and serotonin transporters.
  • CNS-penetrant, suitable for in vivo and in vitro studies.
  • Hydrochloride salt form for improved handling and solubility.
  • High reported purity (99.5%).
  • Supplied in milligram quantities for research applications.
  • Useful for pharmacology, transporter mechanism, and CNS studies.
Catalog No. 50-002-39920 Supplier Medchemexpress LLC Supplier No. HY1071285MG
Ampreloxetine hydrochloride is an orally active, CNS-penetrant inhibitor of the norepinephrine transporter (NET) and the serotonin uptake transporter (SERT) supplied for research use. It is used in vitro and in vivo to investigate monoamine transporter pharmacology and CNS transporter-related mechanisms.
  • Orally active inhibitor of norepinephrine and serotonin transporters.
  • CNS-penetrant, suitable for in vivo and in vitro studies.
  • Hydrochloride salt form for improved handling and solubility.
  • High reported purity (99.5%).
  • Supplied in milligram quantities for research applications.
  • Useful for pharmacology, transporter mechanism, and CNS studies.