Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(3-fluorophenyl)-5-methyl-2(1H)-pyridinone | 848353-85-5 | >98.0% | 100 MG
SDP

Catalog No. 5000232055
Encompass_Preferred

Fluorofenidone is an orally active pyridone small molecule with anti-fibrotic, antioxidant, and anti-inflammatory properties. This listing describes the analytical standard supplied as a 100 mg solid for research and analytical use.

  • analytical standard for laboratory and analytical applications.
  • intended for research use only.
  • cas number 848353-85-5.
  • molecular formula C12H10FNO.
  • molecular weight 203.21 g/mol.
  • purity greater than 98% (supplier varies).
  • pack size 100 mg, solid powder.

Catalog No. 50-002-32055 Supplier Medchemexpress LLC Supplier No. HY121246100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorofenidone is an orally active pyridone small molecule with anti-fibrotic, antioxidant, and anti-inflammatory properties. This listing describes the analytical standard supplied as a 100 mg solid for research and analytical use.

  • analytical standard for laboratory and analytical applications.
  • intended for research use only.
  • cas number 848353-85-5.
  • molecular formula C12H10FNO.
  • molecular weight 203.21 g/mol.
  • purity greater than 98% (supplier varies).
  • pack size 100 mg, solid powder.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.