Learn More
Medchemexpress LLC 1-(3-fluorophenyl)-5-methylpyridin-2(1H)-one | 848353-85-5 | 99.8% | 50 MG
SDP
Catalog No. 5000232058
Encompass_Preferred
Fluorofenidone (AKF-PD) is an orally active small-molecule research compound with reported anti-fibrotic, antioxidant, and anti-inflammatory effects. Supplied as a purified analytical standard suitable for in vitro and in vivo studies and commonly provided as a 50 mg pack.
  • Orally active small molecule with anti-fibrotic and anti-inflammatory activity.
  • Demonstrated cellular effects (e.g., NIH/3T3 IC50 ~2.75 mM) and in vitro activity in lung and cancer cell lines.
  • Shows protective effects in rodent models of lung injury, cholestasis, and fibrosis.
  • High purity suitable for analytical and preclinical research applications.
  • Molecular formula C12H10FNO; molecular weight 203.21 g/mol.
Catalog No. 50-002-32058 Supplier Medchemexpress LLC Supplier No. HY12124650MG
Fluorofenidone (AKF-PD) is an orally active small-molecule research compound with reported anti-fibrotic, antioxidant, and anti-inflammatory effects. Supplied as a purified analytical standard suitable for in vitro and in vivo studies and commonly provided as a 50 mg pack.
  • Orally active small molecule with anti-fibrotic and anti-inflammatory activity.
  • Demonstrated cellular effects (e.g., NIH/3T3 IC50 ~2.75 mM) and in vitro activity in lung and cancer cell lines.
  • Shows protective effects in rodent models of lung injury, cholestasis, and fibrosis.
  • High purity suitable for analytical and preclinical research applications.
  • Molecular formula C12H10FNO; molecular weight 203.21 g/mol.