Learn More
Medchemexpress LLC 1,3-glyceryl dilinoleate | 15818-46-9 | MFCD00056310 | 616.95 g/mol | C39H68O5 | 50 MG
SDP
Catalog No. 5000282520
Encompass_Preferred
1,3-Glyceryl dilinoleate (1,3-dilinolein) is an unsaturated diacylglycerol used as a biochemical reagent in lipid research and assay development. It features linoleic acid (18:2) side chains at the C-1 and C-3 positions of the glycerol backbone, and has molecular formula C39H68O5 and molecular weight 616.95 g/mol.
  • Unsaturated diacylglycerol suitable for lipid research and assays.
  • Contains linoleic acid chains at both C-1 and C-3 positions.
  • Useful for studies of diacylglycerol signaling and lipid metabolism.
  • Available in small milligram quantities for analytical and biochemical use.
  • Store and handle per certificate of analysis and safety documentation.
Catalog No. 50-002-82520 Supplier Medchemexpress LLC Supplier No. HYW33609450MG
1,3-Glyceryl dilinoleate (1,3-dilinolein) is an unsaturated diacylglycerol used as a biochemical reagent in lipid research and assay development. It features linoleic acid (18:2) side chains at the C-1 and C-3 positions of the glycerol backbone, and has molecular formula C39H68O5 and molecular weight 616.95 g/mol.
  • Unsaturated diacylglycerol suitable for lipid research and assays.
  • Contains linoleic acid chains at both C-1 and C-3 positions.
  • Useful for studies of diacylglycerol signaling and lipid metabolism.
  • Available in small milligram quantities for analytical and biochemical use.
  • Store and handle per certificate of analysis and safety documentation.