Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-glyceryl dilinoleate | 15818-46-9 | MFCD00056310 | ≥95.0% | 616.95 g/mol | C39H68O5 | 100 MG
SDP

Catalog No. 5000282091
Encompass_Preferred

1,3-Glyceryl dilinoleate is an unsaturated diacylglycerol used as a laboratory research reagent for studies of lipids, lipid signaling, and metabolism. It is an oil composed of glycerol esterified with two linoleic acid chains and is supplied with analytical-grade purity.

  • Contains two linoleic acid (18:2) chains esterified to glycerol.
  • Suitable for lipid metabolism and signaling studies.
  • Provided as a neat oil for analytical and biochemical applications.
  • Typical purity ≥95.0%.
  • Molecular weight 616.95 g/mol; formula C39H68O5.

Catalog No. 50-002-82091 Supplier Medchemexpress LLC Supplier No. HYW336094100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Glyceryl dilinoleate is an unsaturated diacylglycerol used as a laboratory research reagent for studies of lipids, lipid signaling, and metabolism. It is an oil composed of glycerol esterified with two linoleic acid chains and is supplied with analytical-grade purity.

  • Contains two linoleic acid (18:2) chains esterified to glycerol.
  • Suitable for lipid metabolism and signaling studies.
  • Provided as a neat oil for analytical and biochemical applications.
  • Typical purity ≥95.0%.
  • Molecular weight 616.95 g/mol; formula C39H68O5.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.