Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-glyceryl dilinoleate | 15818-46-9 | MFCD00056310 | ≥95.0% | 616.95 g·mol⁻¹ | C39H68O5 | 25 MG
SDP

Catalog No. 5000280554
Encompass_Preferred

1,3-Glyceryl dilinoleate is an unsaturated diacylglycerol used as a research reagent in lipid chemistry and biochemical studies. It has chemical formula C39H68O5 and a molecular weight of 616.95 g·mol⁻¹, and is commonly supplied in small-mass packages for laboratory use.

  • Used as a reagent in lipid and membrane research.
  • Chemical formula C39H68O5 and molecular weight 616.95 g·mol⁻¹.
  • CAS number 15818-46-9 for unambiguous identification.
  • Commonly supplied in mg-scale quantities for analytical and research use.
  • Typically provided at high purity suitable for research applications.

Catalog No. 50-002-80554 Supplier Medchemexpress LLC Supplier No. HYW33609425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Glyceryl dilinoleate is an unsaturated diacylglycerol used as a research reagent in lipid chemistry and biochemical studies. It has chemical formula C39H68O5 and a molecular weight of 616.95 g·mol⁻¹, and is commonly supplied in small-mass packages for laboratory use.

  • Used as a reagent in lipid and membrane research.
  • Chemical formula C39H68O5 and molecular weight 616.95 g·mol⁻¹.
  • CAS number 15818-46-9 for unambiguous identification.
  • Commonly supplied in mg-scale quantities for analytical and research use.
  • Typically provided at high purity suitable for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.