Learn More
eMolecules​ 1-(3-Hydroxy-2-methylphenyl)ethanone | Combi-Blocks | 69976-81-4 | MFCD13689052 | 150.177 | C9H10O2 | 97.000 | CC(=O)c1cccc(O)c1C | 1g | 517037269
Supplier:  eMolecules​ 517037269
Encompass_Preferred
1-(3-Hydroxy-2-methylphenyl)ethanone | Combi-Blocks | 69976-81-4 | MFCD13689052 | 150.177 | C9H10O2 | 97.000 | CC(=O)c1cccc(O)c1C | 1g | 517037269
Catalog No. NC2228416