Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Iodobenzyl)piperidine, ≥97%, Thermo Scientific™

Catalog No. CC45211DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC45211DE 5 g
CC45211CB 250 mg
CC45211DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC45211DE Supplier Thermo Scientific Chemicals Supplier No. CC45211DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 859850-87-6
Molecular Formula C12H16IN
Molecular Weight (g/mol) 301.171
MDL Number MFCD06797854
InChI Key DCRZUICHKJGHQK-UHFFFAOYSA-N
Synonym 1-3-iodobenzyl piperidine, 1-3-iodophenyl methyl piperidine, piperidine,1-3-iodophenyl methyl, 1-3-iodo-benzyl-piperidine
PubChem CID 17750314
IUPAC Name 1-[(3-iodophenyl)methyl]piperidine
SMILES C1CCN(CC1)CC2=CC(=CC=C2)I

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 301.17
Percent Purity 97%
Chemical Name or Material 1-(3-Iodobenzyl)piperidine
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.