Learn More
Medchemexpress LLC 1-(3-methoxybenzyl)-3-(4-(pyridin-4-yl)thiazol-2-yl)urea | 1342276-76-9 | 98.2% | 340.40 g/mol | C17H16N4O2S | 1 MG
SDP
Catalog No. 5000240934
Encompass_Preferred
RKI-1313 is a small-molecule ROCK kinase inhibitor supplied as a crystalline solid for research use. It selectively inhibits ROCK isoforms with reported IC50 values indicating greater potency for ROCK2, and is used in cell-signaling and kinase inhibition studies. The compound has formula C17H16N4O2S, molecular weight 340.40 g/mol, and is typically supplied at high purity.
Catalog No. 50-002-40934 Supplier Medchemexpress LLC Supplier No. HY1072091MG
RKI-1313 is a small-molecule ROCK kinase inhibitor supplied as a crystalline solid for research use. It selectively inhibits ROCK isoforms with reported IC50 values indicating greater potency for ROCK2, and is used in cell-signaling and kinase inhibition studies. The compound has formula C17H16N4O2S, molecular weight 340.40 g/mol, and is typically supplied at high purity.