Learn More
Medchemexpress LLC 1-(3-Methoxyphenyl)ethanamine | 62409-13-6 | 151.21 | 25 G
SDP
Catalog No. 5000402645
Encompass_Preferred
1-(3-Methoxyphenyl)ethanamine is a drug intermediate used for the synthesis of various active compounds. Its molecular formula is C9H13NO and it has a molecular weight of 151.21.
  • Used for the synthesis of various active compounds.
  • Pure form storage recommendations: -20°C for 3 years, or 4°C for 2 years.
  • In solvent storage recommendations: -80°C for 6 months, or -20°C for 1 month.
  • Intended for laboratory use.
Catalog No. 50-004-02645 Supplier Medchemexpress LLC Supplier No. 25GHY20143
1-(3-Methoxyphenyl)ethanamine is a drug intermediate used for the synthesis of various active compounds. Its molecular formula is C9H13NO and it has a molecular weight of 151.21.
  • Used for the synthesis of various active compounds.
  • Pure form storage recommendations: -20°C for 3 years, or 4°C for 2 years.
  • In solvent storage recommendations: -80°C for 6 months, or -20°C for 1 month.
  • Intended for laboratory use.