Learn More
Medchemexpress LLC 1-[(3-methoxyphenyl)methyl]-N-(4-methyl-3-morpholin-4-ylsulfonylphenyl)-3-(4-methylphenyl)pyrazole | 1609564-75-1 | 99.3% | 560.66 | C30H32N4O5S | 5 MG
SDP
Catalog No. 5000264889
Encompass_Preferred
DY268 is a research compound that functions as a potent farnesoid X receptor (FXR) antagonist used in biochemical and cell-based studies of FXR signaling and drug-induced liver injury. Analytical data and supplier listings report defined composition, high purity, and suitability for in vitro assays.
  • Potent farnesoid X receptor antagonist (IC50 = 7.5 nM).
  • Inhibits FXR transactivation in cell-based assays (IC50 = 468 nM).
  • High reported purity (~99.3% by HPLC).
  • Defined chemical formula C30H32N4O5S and molecular weight 560.66 Da.
  • Soluble in DMSO for in vitro use.
  • Available in small research quantities suitable for assay development.
Catalog No. 50-002-64889 Supplier Medchemexpress LLC Supplier No. HY1102675MG
DY268 is a research compound that functions as a potent farnesoid X receptor (FXR) antagonist used in biochemical and cell-based studies of FXR signaling and drug-induced liver injury. Analytical data and supplier listings report defined composition, high purity, and suitability for in vitro assays.
  • Potent farnesoid X receptor antagonist (IC50 = 7.5 nM).
  • Inhibits FXR transactivation in cell-based assays (IC50 = 468 nM).
  • High reported purity (~99.3% by HPLC).
  • Defined chemical formula C30H32N4O5S and molecular weight 560.66 Da.
  • Soluble in DMSO for in vitro use.
  • Available in small research quantities suitable for assay development.