Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(3-Methyloxetan-3-yl)methanamine

Catalog No. CDS019169 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS019169 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS019169 Supplier Sigma Aldrich Supplier No. CDS019169100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-chloro-2-propanol is formed as an intermediate during the degradation of meso-bis-(1-chloro-2-propyl)ether by rhodococcus sp. strain dtb.1-chloro-2-propanol has been used as:
  • Chemical intermediate for the manufacture of propylene oxide, a starting material for production of polyurethane polyols and propylene glycol
  • As α,α-halohydrin standard during the enzymatic synthesis of α,α-halohydrins from gaseous alkenes

Specifications

Molecular Formula C5H11NO
Linear Formula C5H11NO
MDL Number MFCD08703640
Quantity 25 g
Molecular Weight (g/mol) 101.15
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.