Learn More
Medchemexpress LLC [1-(3-Methylphenyl)benzimidazol-5-yl]-piperidin-1-ylmethanone | 451496-96-1 | MFCD04422127 | 99.8% | 319.4 g/mol | C20H21N3O | 10MM 1ML
SDP
Catalog No. 5000237028
Encompass_Preferred
ML148 is a potent and selective inhibitor of 15-hydroxyprostaglandin dehydrogenase (15-PGDH) used as a research reagent to probe prostaglandin-signaling pathways. The compound is available as a 10 mM solution in DMSO or as solid material in multiple mass sizes. Reported IC50 against 15-PGDH is 56 nM. Molecular formula C20H21N3O; molecular weight 319.4 g/mol; reported purity ~99.8%.
  • Potent and selective 15-PGDH inhibitor, IC50 56 nM.
  • Offered as 10 mM DMSO solution and as solid quantities for flexibility in assays.
  • High purity (~99.8%) suitable for biochemical and cell-based research.
  • Well-characterized chemical identity: C20H21N3O, 319.4 g/mol.
  • Supplied with spectral and analytical data for quality assurance.
Catalog No. 50-002-37028 Supplier Medchemexpress LLC Supplier No. HY12354810MM 1ML
ML148 is a potent and selective inhibitor of 15-hydroxyprostaglandin dehydrogenase (15-PGDH) used as a research reagent to probe prostaglandin-signaling pathways. The compound is available as a 10 mM solution in DMSO or as solid material in multiple mass sizes. Reported IC50 against 15-PGDH is 56 nM. Molecular formula C20H21N3O; molecular weight 319.4 g/mol; reported purity ~99.8%.
  • Potent and selective 15-PGDH inhibitor, IC50 56 nM.
  • Offered as 10 mM DMSO solution and as solid quantities for flexibility in assays.
  • High purity (~99.8%) suitable for biochemical and cell-based research.
  • Well-characterized chemical identity: C20H21N3O, 319.4 g/mol.
  • Supplied with spectral and analytical data for quality assurance.