Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Methylphenyl)ethanol, 95%, Thermo Scientific™

Catalog No. AAH3278403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3278403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3278403 Supplier Thermo Scientific Chemicals Supplier No. H3278403

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 25675-28-9
Molecular Formula C9H12O
Quantity 1 g
MDL Number MFCD00046635
Synonym 1-3-methylphenyl ethanol, 1-3-methylphenyl ethan-1-ol, 1-m-tolyl ethanol, a,3-dimethylbenzenemethanol, alpha-3-dimethylbenzyl alcohol, 1-m-tolyl-ethanol, 1-m-methylphenyl ethanol, m-tolylmethylcarbinol, 1-3-methylphenyl-1-ethanol, +/--1-3-methylphenyl ethanol
InChI Key SPNHUMWMKXWVIU-UHFFFAOYSA-N
SMILES CC1=CC(=CC=C1)C(C)O
IUPAC Name 1-(3-methylphenyl)ethanol
Molecular Weight (g/mol) 136.194
PubChem CID 110952
Formula Weight 136.2
Percent Purity 95%
Chemical Name or Material 1-(3-Methylphenyl)ethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.