Learn More
Medchemexpress LLC 1,3-octadecanediol, 2-amino-, (2S,3R)- | 764-22-7 | MFCD07371371 | 99.6% | 301.51 g/mol | C18H39NO2 | 50 MG
SDP
Catalog No. 5000300331
Encompass_Preferred
D-Erythro-dihydrosphingosine is a sphingoid base (sphinganine) that directly inhibits cytosolic phospholipase A2α (cPLA2α). Supplied as a white to off-white solid for research use, it has molecular formula C18H39NO2, molecular weight 301.51 g/mol, and CAS 764-22-7. The Certificate of Analysis reports an LCMS purity of 99.55% and recommends sealed storage at -20°C (in solvent: -80°C up to 6 months).
  • Direct inhibitor of cPLA2α for mechanistic studies.
  • High purity (99.55%) for reproducible results.
  • Well-characterized chemical identity (C18H39NO2; MW 301.51 g/mol).
  • Solid, white to off-white appearance for easy handling.
  • Recommended storage conditions to maintain compound integrity.
Catalog No. 50-003-00331 Supplier Medchemexpress LLC Supplier No. HYW01983850MG
D-Erythro-dihydrosphingosine is a sphingoid base (sphinganine) that directly inhibits cytosolic phospholipase A2α (cPLA2α). Supplied as a white to off-white solid for research use, it has molecular formula C18H39NO2, molecular weight 301.51 g/mol, and CAS 764-22-7. The Certificate of Analysis reports an LCMS purity of 99.55% and recommends sealed storage at -20°C (in solvent: -80°C up to 6 months).
  • Direct inhibitor of cPLA2α for mechanistic studies.
  • High purity (99.55%) for reproducible results.
  • Well-characterized chemical identity (C18H39NO2; MW 301.51 g/mol).
  • Solid, white to off-white appearance for easy handling.
  • Recommended storage conditions to maintain compound integrity.