Learn More
Medchemexpress LLC 1,3-Oxazole | 288-42-6 | 100.0% | 69.06 | 5 G
SDP
Catalog No. 5000403836
Encompass_Preferred
1,3-Oxazole is a biochemical reagent that can be used as a biological material or organic compound for life science related research. Oxazole and its derivatives are actively involved as building blocks for biochemicals and pharmaceuticals like flopristin and darglitazone. It is also involved in industrial applications such as in dyes, fluorescent brightening agents, textile auxiliaries and plastics.
  • Used as a biological material or organic compound for life science research
  • Acts as building blocks for biochemicals and pharmaceuticals
  • Applied in industrial applications including dyes
  • Applied in fluorescent brightening agents
  • Applied in textile auxiliaries
  • Applied in plastics
Catalog No. 50-004-03836 Supplier Medchemexpress LLC Supplier No. HY346315G
1,3-Oxazole is a biochemical reagent that can be used as a biological material or organic compound for life science related research. Oxazole and its derivatives are actively involved as building blocks for biochemicals and pharmaceuticals like flopristin and darglitazone. It is also involved in industrial applications such as in dyes, fluorescent brightening agents, textile auxiliaries and plastics.
  • Used as a biological material or organic compound for life science research
  • Acts as building blocks for biochemicals and pharmaceuticals
  • Applied in industrial applications including dyes
  • Applied in fluorescent brightening agents
  • Applied in textile auxiliaries
  • Applied in plastics