Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-oxazolidine-2-thione | 5840-81-3 | MFCD00125763 | ≥98.0% | 103.15 g/mol | C3H5NOS | 10 MG
SDP

Catalog No. 5000300390
Encompass_Preferred

1,3-oxazolidine-2-thione (CAS 5840-81-3) is a small heterocyclic organosulfur compound supplied for laboratory research and reference use. It is typically a white to off-white solid with molecular formula C3H5NOS and molecular weight about 103.15 g/mol. The substance has been used in biological studies and is provided with specified purity for experimental applications.

  • Small heterocyclic organosulfur compound
  • White to off-white solid appearance
  • Molecular formula C3H5NOS and molecular weight 103.15 g/mol
  • Soluble in DMSO (common laboratory solvent)
  • Supplied with reported purity ≥98.0%
  • Intended for laboratory research and reference use only

Catalog No. 50-003-00390 Supplier Medchemexpress LLC Supplier No. HYW03898510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-oxazolidine-2-thione (CAS 5840-81-3) is a small heterocyclic organosulfur compound supplied for laboratory research and reference use. It is typically a white to off-white solid with molecular formula C3H5NOS and molecular weight about 103.15 g/mol. The substance has been used in biological studies and is provided with specified purity for experimental applications.

  • Small heterocyclic organosulfur compound
  • White to off-white solid appearance
  • Molecular formula C3H5NOS and molecular weight 103.15 g/mol
  • Soluble in DMSO (common laboratory solvent)
  • Supplied with reported purity ≥98.0%
  • Intended for laboratory research and reference use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.