Learn More
Medchemexpress LLC 1,3-oxazolidine-2-thione | 5840-81-3 | MFCD00125763 | 99.4% | 103.14 g·mol⁻¹ | C3H5NOS | 25 MG
SDP
Catalog No. 5000303924
Encompass_Preferred
1,3-Oxazolidine-2-thione (CAS 5840-81-3) is a small-molecule oxazolidinethione used as a research reagent. It is a low-molecular-weight compound (C3H5NOS, 103.14 g·mol⁻¹) supplied at high purity for laboratory studies. It has been reported to increase thyroid size and to markedly depress hepatic trimethylamine oxidase activity in avian models. For research use only.
  • High purity suitable for analytical and biological research.
  • Well-characterized molecular properties for reproducible experiments.
  • Available in small pack sizes for screening and mechanistic studies.
  • Reported biological activity relevant to endocrine and hepatic research.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-003-03924 Supplier Medchemexpress LLC Supplier No. 25MGHYW038985
1,3-Oxazolidine-2-thione (CAS 5840-81-3) is a small-molecule oxazolidinethione used as a research reagent. It is a low-molecular-weight compound (C3H5NOS, 103.14 g·mol⁻¹) supplied at high purity for laboratory studies. It has been reported to increase thyroid size and to markedly depress hepatic trimethylamine oxidase activity in avian models. For research use only.
  • High purity suitable for analytical and biological research.
  • Well-characterized molecular properties for reproducible experiments.
  • Available in small pack sizes for screening and mechanistic studies.
  • Reported biological activity relevant to endocrine and hepatic research.
  • Intended for research use only; not for human or veterinary use.