Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-oxazolidine-2-thione | 5840-81-3 | MFCD00125763 | 99.4% | 103.14 g/mol | C3H5NOS | 5 MG
SDP

Catalog No. 5000299809
Encompass_Preferred

1,3-Oxazolidine-2-thione is a small heterocyclic thiocarbonyl compound supplied for laboratory research. It is provided as a solid research reagent with documented physical and storage properties and reported biological effects in animal studies (effects on thyroid size and hepatic trimethylamine oxidase activity). The material is characterized by a defined purity, molecular formula, and high solubility in DMSO.

  • Small heterocyclic thiocarbonyl solid.
  • Purity 99.44%.
  • Molecular weight 103.14 g/mol; formula C3H5NOS.
  • High DMSO solubility: 250 mg/mL (ultrasonic assistance recommended).
  • Powder storage: -20°C up to 3 years, 4°C up to 2 years.
  • Reported biological activity affecting thyroid size and hepatic trimethylamine oxidase in animal studies.
  • Available in multiple small pack sizes for research use.

Catalog No. 50-002-99809 Supplier Medchemexpress LLC Supplier No. HYW0389855MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Oxazolidine-2-thione is a small heterocyclic thiocarbonyl compound supplied for laboratory research. It is provided as a solid research reagent with documented physical and storage properties and reported biological effects in animal studies (effects on thyroid size and hepatic trimethylamine oxidase activity). The material is characterized by a defined purity, molecular formula, and high solubility in DMSO.

  • Small heterocyclic thiocarbonyl solid.
  • Purity 99.44%.
  • Molecular weight 103.14 g/mol; formula C3H5NOS.
  • High DMSO solubility: 250 mg/mL (ultrasonic assistance recommended).
  • Powder storage: -20°C up to 3 years, 4°C up to 2 years.
  • Reported biological activity affecting thyroid size and hepatic trimethylamine oxidase in animal studies.
  • Available in multiple small pack sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.