Learn More
Cayman Chemical 1,3-PBIT (dihydrobromide), Purity: ≥98%, CAS: 200716-66-1, 10 mg
SDP
Catalog No. 502896779
Encompass_Preferred
A potent inhibitor of iNOS with a Ki value of 47 nM (Ki values for eNOS and nNOS are 9 and 25 µM, respectively)
Catalog No. 50-289-6779 Supplier Cayman Chemical Supplier No. 8150010 MG
A potent inhibitor of iNOS with a Ki value of 47 nM (Ki values for eNOS and nNOS are 9 and 25 µM, respectively)