Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Pentadiene (cis- and trans- mixture) (stabilized with TBC), TCI America™
SDP

Catalog No. P1841100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1841100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1841100ML Supplier TCI America Supplier No. P1841100ML

missing translation for 'validMainframeMsg'

Specifications

CAS 504-60-9
Color Yellow
Boiling Point 42°C
Molecular Formula C5H8
MDL Number MFCD00009295
UN Number 3295
Quantity 100 mL
Synonym 1,3-pentadiene, trans-1,3-pentadiene, piperylene, trans-piperylene, e-1,3-pentadiene, 1-methylbutadiene, 3e-penta-1,3-diene, trans-1-methylbutadiene, 1,3-pentadiene, 3e, trans-penta-1,3-diene
InChI Key PMJHHCWVYXUKFD-SNAWJCMRSA-N
SMILES CC=CC=C
IUPAC Name (3E)-penta-1,3-diene
Molecular Weight (g/mol) 68.119
PubChem CID 62204
ChEBI CHEBI:74165
Formula Weight 68.12
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Pentadiene (cis- and trans- mixture) (stabilized with TBC)
Show More Show Less
EINECSNumber (2)-0020
RTECSNumber RZ2464000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.