Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Phenylene Diisocyanate 98.0+%, TCI America™
SDP

Catalog No. P10921G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10921G 1 g
P10925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10921G Supplier TCI America Supplier No. P10921G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 123-61-5
Molecular Formula C8H4N2O2
Molecular Weight (g/mol) 160.132
MDL Number MFCD00002018
InChI Key VGHSXKTVMPXHNG-UHFFFAOYSA-N
Synonym 1,3-phenylene diisocyanate, m-phenylene diisocyanate, benzene, 1,3-diisocyanato, benzene 1,3-diisocyanate, nacconate 400, benzene, m-diisocyanato, m-phenylene isocyanate, benzene-1,3-diisocyanate, isocyanic acid, m-phenylene ester, unii-b44h9mfh1s
PubChem CID 31262
IUPAC Name 1,3-diisocyanatobenzene
SMILES C1=CC(=CC(=C1)N=C=O)N=C=O

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 103°C
Quantity 1 g
UN Number 2811
Formula Weight 160.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Phenylene Diisocyanate
EINECSNumber (3)-2215
RTECSNumber NR0150000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.