Learn More
Medchemexpress LLC 1,3-phenylene tetraphenyl bis(phosphate) | 57583-54-7 | MFCD01755688 | 574.45 g·mol⁻¹ | C30H24O8P2 | 50 MG
SDP
Catalog No. 5000285191
Encompass_Preferred
1,3-Phenylene tetraphenyl bis(phosphate) is an aryl bis(phosphate) compound used as a flame retardant and research reagent. Supplied in small laboratory quantities, it is intended for materials research and formulation testing where evaluation of thermal stability and fire resistance in polymer systems is required.
  • Aryl bis(phosphate) flame retardant for polymer additive studies.
  • Supplied in laboratory-scale quantities suitable for formulation testing.
  • High molecular weight and aromatic structure that support thermal stability.
  • Applicable to evaluation in thermoplastics and thermosets.
  • Datasheet and certificate of analysis available on request.
Catalog No. 50-002-85191 Supplier Medchemexpress LLC Supplier No. HYW39212250MG
1,3-Phenylene tetraphenyl bis(phosphate) is an aryl bis(phosphate) compound used as a flame retardant and research reagent. Supplied in small laboratory quantities, it is intended for materials research and formulation testing where evaluation of thermal stability and fire resistance in polymer systems is required.
  • Aryl bis(phosphate) flame retardant for polymer additive studies.
  • Supplied in laboratory-scale quantities suitable for formulation testing.
  • High molecular weight and aromatic structure that support thermal stability.
  • Applicable to evaluation in thermoplastics and thermosets.
  • Datasheet and certificate of analysis available on request.