Learn More
Medchemexpress LLC 1,3-phenylene tetraphenyl bis(phosphate) | 57583-54-7 | MFCD01755688 | >98.0% | 574.45 g/mol | C30H24O8P2 | 5 MG
SDP
Catalog No. 5000285228
Encompass_Preferred
1,3-Phenylene tetraphenyl bis(phosphate) is an aryl bis(phosphate) compound used as a flame retardant and a specialty research reagent. It is commonly used in flame retardant formulations, materials research, and as an analytical reference standard for organophosphate studies.
  • Used as a flame retardant and additive in materials research.
  • Aryl bis(phosphate) chemical class with organophosphate functionality.
  • High chemical purity suitable for analytical and research use.
  • Well characterized: CAS 57583-54-7, formula C30H24O8P2, MW 574.45 g/mol.
  • Available in small research quantities (e.g., 5 MG).
Catalog No. 50-002-85228 Supplier Medchemexpress LLC Supplier No. HYW3921225MG
1,3-Phenylene tetraphenyl bis(phosphate) is an aryl bis(phosphate) compound used as a flame retardant and a specialty research reagent. It is commonly used in flame retardant formulations, materials research, and as an analytical reference standard for organophosphate studies.
  • Used as a flame retardant and additive in materials research.
  • Aryl bis(phosphate) chemical class with organophosphate functionality.
  • High chemical purity suitable for analytical and research use.
  • Well characterized: CAS 57583-54-7, formula C30H24O8P2, MW 574.45 g/mol.
  • Available in small research quantities (e.g., 5 MG).