Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Phenylenediamine Dihydrochloride 99.0+%, TCI America™
SDP

Catalog No. P0172500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0172500G 500 g
P017225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0172500G Supplier TCI America Supplier No. P0172500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 541-69-5
Molecular Formula C6H10Cl2N2
Molecular Weight (g/mol) 181.06
MDL Number MFCD00012975
InChI Key SVTOYMIYCMHPIV-UHFFFAOYSA-N
Synonym benzene-1,3-diamine dihydrochloride, m-phenylenediamine dihydrochloride, 1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine dihydrochloride, 1,3-benzenediamine, dihydrochloride, m-phenylenediamine hydrochloride, unii-se5rlo93tg, 1,3-diaminobenzene dihydrochloride, m-aminoaniline dihydrochloride, 3-aminoaniline dihydrochloride
PubChem CID 10941
IUPAC Name benzene-1,3-diamine;dihydrochloride
SMILES C1=CC(=CC(=C1)N)N.Cl.Cl

Specifications

Color Purple-White
Quantity 500 g
UN Number 2811
Formula Weight 181.06
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Phenylenediamine Dihydrochloride
EINECSNumber (3)-0185
RTECSNumber SS9800000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.