Learn More
Medchemexpress LLC 1,3-Propanediol, 2,2'-(1,3-propanediyldiimino)bis[2-(hydroxymethyl)- | 64431-96-5 | 99.9% | 282.33 | 1 ML
SDP
Catalog No. 5000438030
Encompass_Preferred
Bis-tris propane (BTP) is a water-soluble buffer substance. It can be used as a suitable buffer for polymerase chain reaction (PCR) and can enhance the stability or activity of restriction enzymes. In PCR reactions, Bis-tris propane buffer demonstrates high fidelity by stabilizing the pH of the reaction system and reducing errors introduced by DNA polymerase.
  • Water-soluble buffer substance.
  • Suitable buffer for polymerase chain reaction (PCR).
  • Enhances the stability or activity of restriction enzymes.
  • Exhibits high fidelity in PCR reactions by stabilizing pH and reducing DNA polymerase errors.
Catalog No. 50-004-38030 Supplier Medchemexpress LLC Supplier No. HY12639910MM/1ML
Add to Cart
Add to Cart
Bis-tris propane (BTP) is a water-soluble buffer substance. It can be used as a suitable buffer for polymerase chain reaction (PCR) and can enhance the stability or activity of restriction enzymes. In PCR reactions, Bis-tris propane buffer demonstrates high fidelity by stabilizing the pH of the reaction system and reducing errors introduced by DNA polymerase.
  • Water-soluble buffer substance.
  • Suitable buffer for polymerase chain reaction (PCR).
  • Enhances the stability or activity of restriction enzymes.
  • Exhibits high fidelity in PCR reactions by stabilizing pH and reducing DNA polymerase errors.