Learn More
Medchemexpress LLC 1,3-propanediol, 2-amino-2-(1-nonyl-1H-1,2,3-triazol-4-yl) hydrochloride | 2376132-24-8 | 99.9% | 320.86 | C14H29ClN4O2 | 10 MG
SDP
Catalog No. 5000295115
Encompass_Preferred
KARI 201 hydrochloride is a dual-action small-molecule research compound that functions as a selective, brain-penetrant acid sphingomyelinase (ASM) inhibitor and a ghrelin receptor (GHSR) agonist. It is intended for preclinical in vitro and in vivo studies and is supplied as a solid for laboratory research use only.
  • Selective acid sphingomyelinase inhibitor.
  • Ghrelin receptor agonist with brain penetration.
  • High purity, 99.9% (HPLC).
  • Molecular weight 320.86 g·mol-1.
  • Supplied as a 10 mg solid; store sealed at -20°C.
  • For research use only; not for human or diagnostic use.
Catalog No. 50-002-95115 Supplier Medchemexpress LLC Supplier No. HY171962A10MG
KARI 201 hydrochloride is a dual-action small-molecule research compound that functions as a selective, brain-penetrant acid sphingomyelinase (ASM) inhibitor and a ghrelin receptor (GHSR) agonist. It is intended for preclinical in vitro and in vivo studies and is supplied as a solid for laboratory research use only.
  • Selective acid sphingomyelinase inhibitor.
  • Ghrelin receptor agonist with brain penetration.
  • High purity, 99.9% (HPLC).
  • Molecular weight 320.86 g·mol-1.
  • Supplied as a 10 mg solid; store sealed at -20°C.
  • For research use only; not for human or diagnostic use.