Learn More
Medchemexpress LLC 1,3-Propanediol, 2-bromo-2-nitro- | 52-51-7 | 98.0% | 25 G
SDP
Catalog No. 5000414350
Encompass_Preferred
Bronopol is an antibacterial agent with low toxicity to mammals and high activity, particularly against Gram-negative bacteria. It functions by oxidizing protein thiols, which inhibits enzymatic activity and thereby exhibits its antibacterial properties. Bronopol also acts as a formaldehyde releaser. It is intended for research use only and not for sale to patients.
  • Antibacterial agent: Effective against bacteria, especially gram-negative strains.
  • Mechanism of action: Oxidizes protein thiols and inhibits enzymatic activity.
  • Formaldehyde releaser: Contributes to its biological activity.
  • Low toxicity to mammals.
Catalog No. 50-004-14350 Supplier Medchemexpress LLC Supplier No. 25GHYB1217
Bronopol is an antibacterial agent with low toxicity to mammals and high activity, particularly against Gram-negative bacteria. It functions by oxidizing protein thiols, which inhibits enzymatic activity and thereby exhibits its antibacterial properties. Bronopol also acts as a formaldehyde releaser. It is intended for research use only and not for sale to patients.
  • Antibacterial agent: Effective against bacteria, especially gram-negative strains.
  • Mechanism of action: Oxidizes protein thiols and inhibits enzymatic activity.
  • Formaldehyde releaser: Contributes to its biological activity.
  • Low toxicity to mammals.