Learn More
Medchemexpress LLC 1,3-Propanediol, 2-bromo-2-nitro- | 52-51-7 | 98.0% | 50 G
SDP
Catalog No. 5000414075
Encompass_Preferred
Bronopol is an antibacterial agent with low toxicity to mammals and high activity, especially against Gram-negative bacteria. It oxidizes protein thiols, inhibits enzymatic activity, and exhibits antibacterial activity. Bronopol is also a formaldehyde releaser. It is for research use only and not sold to patients.
  • Antibacterial agent
  • Low toxicity to mammals
  • High activity against gram-negative bacteria
  • Oxidizes protein thiols
  • Inhibits enzymatic activity
  • Formaldehyde releaser
  • Solid appearance
  • White to off-white color
Catalog No. 50-004-14075 Supplier Medchemexpress LLC Supplier No. HYB121750G
Bronopol is an antibacterial agent with low toxicity to mammals and high activity, especially against Gram-negative bacteria. It oxidizes protein thiols, inhibits enzymatic activity, and exhibits antibacterial activity. Bronopol is also a formaldehyde releaser. It is for research use only and not sold to patients.
  • Antibacterial agent
  • Low toxicity to mammals
  • High activity against gram-negative bacteria
  • Oxidizes protein thiols
  • Inhibits enzymatic activity
  • Formaldehyde releaser
  • Solid appearance
  • White to off-white color