Learn More
Medchemexpress LLC 1,3-Propanediol | 504-63-2 | MFCD00002949 | 100.0% | 76.09 | C3H8O2 | 1 ML
SDP
Catalog No. 5000300905
Encompass_Preferred
1,3-Propanediol is a small, water-miscible diol (C3H8O2, MW 76.09, CAS 504-63-2) produced by microbial fermentation of glycerol. It is supplied for research and analytical use in both solution and bulk formats, and is offered as a reference standard for analytical applications.
  • Suitable as an analytical reference standard.
  • Provided in ready-to-use 10 mM DMSO solution for easy dilution.
  • Available in multiple pack sizes to support small-scale and bulk workflows.
  • High purity grades listed to support accurate analytical and research results.
  • Compatible with common solvents and analytical methods.
Catalog No. 50-003-00905 Supplier Medchemexpress LLC Supplier No. HYW01775810MM/1ML
1,3-Propanediol is a small, water-miscible diol (C3H8O2, MW 76.09, CAS 504-63-2) produced by microbial fermentation of glycerol. It is supplied for research and analytical use in both solution and bulk formats, and is offered as a reference standard for analytical applications.
  • Suitable as an analytical reference standard.
  • Provided in ready-to-use 10 mM DMSO solution for easy dilution.
  • Available in multiple pack sizes to support small-scale and bulk workflows.
  • High purity grades listed to support accurate analytical and research results.
  • Compatible with common solvents and analytical methods.