Learn More
Medchemexpress LLC 1,3-propanediol | 504-63-2 | MFCD00002949 | 100.0% | 76.09 g/mol | C3H8O2 | 25 G
SDP
Catalog No. 5000302802
Encompass_Preferred
1,3-Propanediol is a colorless, viscous, water-miscible diol used as a chemical intermediate, analytical standard, and formulation ingredient for laboratory research. It is intended for research and analytical applications and is provided with safety and analytical documentation to support laboratory use.
  • High purity with GC-verified certificate of analysis.
  • Chemical formula C3H8O2 and molecular weight 76.09 g/mol.
  • Suitable as an analytical standard and reagent for polymer synthesis, formulation, and analytical workflows.
  • Miscible with water and common organic solvents, enabling versatile sample preparation.
  • Accompanied by safety data sheet and certificate of analysis for traceability.
Catalog No. 50-003-02802 Supplier Medchemexpress LLC Supplier No. 25GHYW017758
1,3-Propanediol is a colorless, viscous, water-miscible diol used as a chemical intermediate, analytical standard, and formulation ingredient for laboratory research. It is intended for research and analytical applications and is provided with safety and analytical documentation to support laboratory use.
  • High purity with GC-verified certificate of analysis.
  • Chemical formula C3H8O2 and molecular weight 76.09 g/mol.
  • Suitable as an analytical standard and reagent for polymer synthesis, formulation, and analytical workflows.
  • Miscible with water and common organic solvents, enabling versatile sample preparation.
  • Accompanied by safety data sheet and certificate of analysis for traceability.