Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-propanediol | 504-63-2 | MFCD00002949 | 98.8% | 76.09 g/mol | C3H8O2 | 10 G
SDP

Catalog No. 5000301662
Encompass_Preferred

1,3-propanediol is a small diol used as an analytical standard, solvent, and research reagent. It occurs naturally via glycerol fermentation and is supplied for analytical and synthetic laboratory applications.

  • High purity suitable for analytical applications.
  • Liquid form; density 1.053 g/cm³.
  • Colorless to light yellow appearance.
  • Molecular formula C3H8O2; molecular weight 76.09 g/mol.
  • CAS number 504-63-2 for unambiguous identification.
  • Available in multiple pack sizes, including solutions and solid packs for lab use.

Catalog No. 50-003-01662 Supplier Medchemexpress LLC Supplier No. HYW01775810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-propanediol is a small diol used as an analytical standard, solvent, and research reagent. It occurs naturally via glycerol fermentation and is supplied for analytical and synthetic laboratory applications.

  • High purity suitable for analytical applications.
  • Liquid form; density 1.053 g/cm³.
  • Colorless to light yellow appearance.
  • Molecular formula C3H8O2; molecular weight 76.09 g/mol.
  • CAS number 504-63-2 for unambiguous identification.
  • Available in multiple pack sizes, including solutions and solid packs for lab use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.