Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-Propanediol | 504-63-2 | MFCD00002949 | 99.97% | 76.09 g/mol | C3H8O2 | 5 G
SDP

Catalog No. 5000301464
Encompass_Preferred

1,3-Propanediol is a small, linear diol used as a reagent, solvent, and reference standard in research and analytical applications. It occurs naturally via glycerol fermentation and is provided at high purity for biochemical studies, polymer synthesis, and formulation work. Solutions in DMSO are available for ready-to-use assays.

  • Used as a reagent and solvent in synthesis and analysis.
  • Occurs naturally via glycerol fermentation.
  • High purity, reported up to 99.97%.
  • Molecular formula C3H8O2; molecular weight 76.09 g/mol.
  • Soluble in DMSO; 10 mM solutions available.
  • Supplied in multiple package sizes for research use.

Catalog No. 50-003-01464 Supplier Medchemexpress LLC Supplier No. HYW0177585G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Propanediol is a small, linear diol used as a reagent, solvent, and reference standard in research and analytical applications. It occurs naturally via glycerol fermentation and is provided at high purity for biochemical studies, polymer synthesis, and formulation work. Solutions in DMSO are available for ready-to-use assays.

  • Used as a reagent and solvent in synthesis and analysis.
  • Occurs naturally via glycerol fermentation.
  • High purity, reported up to 99.97%.
  • Molecular formula C3H8O2; molecular weight 76.09 g/mol.
  • Soluble in DMSO; 10 mM solutions available.
  • Supplied in multiple package sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.