Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Propanediol, 98%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
10L
Packaging:
Glass bottle
Metal drum
504-63-2
C3H8O2
76.09
YPFDHNVEDLHUCE-UHFFFAOYSA-N
1,3-propanediol,trimethylene glycol,1,3-dihydroxypropane,2-deoxyglycerol,1,3-propylene glycol,1,3-propylenediol,1,3-propandiol,2-hydroxymethyl ethanol,omega-propanediol,beta-propylene glycol
10442
CHEBI:16109
propane-1,3-diol
C(CO)CO
504-63-2
C3H8O2
76.09
YPFDHNVEDLHUCE-UHFFFAOYSA-N
1,3-propanediol,trimethylene glycol,1,3-dihydroxypropane,2-deoxyglycerol,1,3-propylene glycol,1,3-propylenediol,1,3-propandiol,2-hydroxymethyl ethanol,omega-propanediol,beta-propylene glycol
10442
CHEBI:16109
propane-1,3-diol
C(CO)CO
Viscosity 52.7 mPa.s (20°C)
Packaging Glass bottle
Boiling Point 214.0°C
Color Undesignated
Density 1.0520g/mL
Flash Point 140°C
Specific Gravity 1.052
Water 0.2% Max. (K.F.)
Refractive Index 1.4380 to 1.4400
Melting Point -32.0°C
Quantity 250mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula HO(CH2)3OH
Beilstein 01,475
Fieser 13,268
Merck Index 15,9885
Solubility Information Solubility in water: 100g/L water. Other solubilities: soluble in ethanol, diethyl ether,, slightly soluble in benzene
Formula Weight 76.09
Physical Form Oily Liquid
Percent Purity 98%
Chemical Name or Material 1, 3-Propanediol
Show More Show Less